Iron(III) oxide, cosmetic, Alfa Aesar™, NanoArc™
Nanoarc Cosmetic Iron Oxide Is An Auburn Material offering Unique Cosmetic Qualitites. The Small Particle Size Results in a Strong Color Development Combined With High Translucency, For a Unique Glowing Look
Manufacturer:  Alfa Aesar 4489514
 View more versions of this product
Catalog No. AA4489514
Description
Description
S.A. 30-60m2/g
Specifications
Specifications
Iron(III) oxide, cosmetic
1309-37-1
MFCD00011008
159.69
Odorless
25g
Sold in collaboration with Nanophase Technologies Corporation
Fe2O3
Red-brown powder
Powder
Red to Brown
Chemical Identifiers
1309-37-1
Fe2O3
MFCD00011008
Safety and Handling
Safety and Handling
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECSNumber : 215-168-2
SDS Documents