missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iron(III) oxide, 98% (metals basis), Alfa Aesar™
Manufacturer: Alfa Aesar 12375C4
Specifications
Iron(III) oxide | |
98% | |
MFCD00011008 | |
159.69 | |
Red to Brown |
1309-37-1 | |
Fe2O3 | |
Red-brown powder | |
Powder | |
25kg |
Chemical Identifiers
1309-37-1 | |
Fe2O3 | |
MFCD00011008 |
Safety and Handling
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECSNumber : 215-168-2