missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(III) oxide, 99.999%, (trace metal basis), -100 mesh, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 193260500

 View more versions of this product

Catalog No. AC193260500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Iron(III) oxide
Trace Metal Basis
100.00%
Fe2O3
Ferric oxide
Glass bottle
Rust or reddish-brown to brown
1309-37-1
Powder
Fe2O3
159.69
5.24g/mL
0.001% max.
50g

Chemical Identifiers

1309-37-1
Fe2O3
159.69
Iron(III) oxide
100.00%
Fe2O3
Ferric oxide
Trace Metal Basis