Please login to your online account to display your discounted pricing

Iron(III) oxide, 99.999%, (trace metal basis), -100 mesh, ACROS Organics™

$192.37 - $1,855.30

Chemical Identifiers

Chemical Name or Material Iron(III) oxide
CAS 1309-37-1
Purity Grade Trace Metal Basis
Assay Percent Range 100.00%
Molecular Formula Fe2O3
Linear Formula Fe2O3
Formula Weight 159.69
Synonym Ferric oxide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC193260500

  SDS
fisher scientific acros
193260500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC193260100

  SDS
fisher scientific acros
193260100
Glass bottle 10g Each for $192.37

AC193262500

  SDS
thermo fisher scientific
AC193262500
Glass bottle 250g Each for $1,855.30
Description & Specifications

Specifications

Chemical Name or Material Iron(III) oxide
CAS 1309-37-1
Purity Grade Trace Metal Basis
Physical Form Powder
Assay Percent Range 100.00%
Molecular Formula Fe2O3
Linear Formula Fe2O3
Formula Weight 159.69
Synonym Ferric oxide
Density 5.24g/mL
Packaging Glass bottle
Total Trace Metal Impurities 0.001% max.
Color Rust or reddish-brown to brown
Quantity 50g

for ultra trace analysis