Please login to your online account to display your discounted pricing

Iron(III) oxide, 99.99% (metals basis), Alfa Aesar™

$377.19 - $377.19

Chemical Identifiers

Chemical Name or Material Iron(III) oxide
CAS 1309-37-1
Physical Form Powder
Assay Percent Range 99.99% (Metal basis)
Molecular Formula Fe2O3
Formula Weight 159.69g/mol
MDL Number MFCD00011008
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1259322

 
alfa aesar
12593-22
100g Each for $377.19

AA1259336

 
alfa aesar
12593-36
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Appearance Red-brown powder
Density 5.24g/cm3
Odor Odorless
Color Red-brown
Quantity 100g
Chemical Name or Material Iron(III) oxide
CAS 1309-37-1
Physical Form Powder
Assay Percent Range 99.99% (Metal basis)
Molecular Formula Fe2O3
Formula Weight 159.69g/mol
MDL Number MFCD00011008
Health Hazard 1 H315-H319-H335 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECS Number 215-168-2