Learn More
Iron(III) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04724730
Description
Iron(III) oxide acts as a pigment in paints, coatings and colored concretes, used in thermalite. Iron oxide is a low cost, nontoxic, environmental friendly catalyst for various reactions such as oxidation of carbon monoxide, reduction of nitrogen oxides and sulfur dioxide by CO, photocatalytic splitting of water, Fischer-Tropsch synthesis of hydrocarbons, catalytic decomposition of industrial dyes, water-gas shift reactions, nucleation of diamonds, conversion of phenolic and other environmentally harmful aromatic compounds, dehydrogenation of ethylbenzene to styrene, and catalytic oxidations of various organic compounds. It is used in magnetic storage devices and in the production of iron, steel and alloys. It functions as a catalyst in the chemical oxidation of hydrogen peroxide used for the treatment of contaminated water with hydrogen peroxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Iron(III) oxide | |
1309-37-1 | |
250 g | |
Fe2O3 | |
14,4028 | |
JEIPFZHSYJVQDO-UHFFFAOYSA-N | |
trioxodiiron | |
159.69 | |
Odorless |
1565°C | |
Powder | |
(metals basis) | |
MFCD00011008 | |
Insoluble in water. Soluble in dilute acids. | |
[Fe][Fe](=O)(=O)=O | |
159.69 | |
99.9% |
Chemical Identifiers
1309-37-1 | |
159.69 | |
JEIPFZHSYJVQDO-UHFFFAOYSA-N | |
[Fe][Fe](=O)(=O)=O |
Fe2O3 | |
MFCD00011008 | |
trioxodiiron |
Safety and Handling
EINECSNumber : 215-168-2
RTECSNumber : NO7400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only