missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(III) oxide, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 47247A1

 View more versions of this product

Catalog No. AA47247A1



Specifications

Specifications

Iron(III) oxide
(metals basis)
14,4028
Powder
Odorless
1kg
1309-37-1
MFCD00011008
159.69
99.9%
1565°C

Chemical Identifiers

1309-37-1
MFCD00011008
Safety and Handling

Safety and Handling

RTECSNumber : NO7400000

TSCA : TSCA

SDS Documents