Please login to your online account to display your discounted pricing

Iron (III) Oxide, 99.9%, metal basis, Alfa Aesar

$303.19 - $303.19

Chemical Identifiers

Chemical Name or Material Iron(III) oxide
CAS 1309-37-1
Physical Form Powder
Assay Percent Range 99.9% (Metal basis)
Molecular Formula Fe2O3
Formula Weight 159.69g/mol
MDL Number MFCD00011008
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1468030

 
alfa aesar
14680-30
250g Each for $303.19
Description & Specifications

Specifications

Appearance Red-brown powder
Density 5.24g/cm3
Color Red-brown
Quantity 250g
Chemical Name or Material Iron(III) oxide
CAS 1309-37-1
Physical Form Powder
Assay Percent Range 99.9% (Metal basis)
Molecular Formula Fe2O3
Formula Weight 159.69g/mol
MDL Number MFCD00011008
Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECS Number 215-168-2
Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.