Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(III) oxalate hexahydrate

Chemical Identifiers

Chemical Name or Material Iron(III) oxalate hexahydrate
CAS 166897-40-1
Formula Weight 483.84 (375.55 Anhydrous)
MDL Number MFCD00054430
UN Number UN3288
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3111606

  SDS
alfa aesar™
3111606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3111622

  SDS
alfa aesar™
3111622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3111614

  SDS
alfa aesar™
3111614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 483.84 (375.55 Anhydrous)
Physical Form Powder
Quantity 5g
Health Hazard 1 Warning
TSCA TSCA
CAS 166897-40-1
UN Number UN3288
Chemical Name or Material Iron(III) oxalate hexahydrate
MDL Number MFCD00054430
Health Hazard 3 GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Safety and Handling

Warning

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.