Please login to your online account to display your discounted pricing

Iron(III) nitrate nonahydrate, 98+% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Iron(III) nitrate nonahydrate
CAS 7782-61-8
Molecular Formula FeN3O9·9H2O
Linear Formula Fe(NO3)3·9H2O
Formula Weight 403.99 (241.86 Anhydrous)
MDL Number MFCD00149708
UN Number UN1466
Merck Index 14,4027
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA12226A1

  SDS
alfa aesar
12226A1
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline, Contains Excess Hno3
Formula Weight 403.99 (241.86 Anhydrous)
Quantity 1kg
CAS 7782-61-8
Density 1.684
Chemical Name or Material Iron(III) nitrate nonahydrate
TSCA TSCA
Odor Acidic
Assay ≥98%
Sensitivity Moisture Sensitive
Molecular Formula FeN3O9·9H2O
Linear Formula Fe(NO3)3·9H2O
MDL Number MFCD00149708
UN Number UN1466
Merck Index 14,4027
Boiling Point 125°C (dec.)
Color Pale Purple
Melting Point 47°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272-H315-H319;
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P221-P210-P220-P280-P305+P351+P338-P501a;
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: FERRIC NITRATE
RTECS Number NO7175000
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P280-P305+P351+P338-P501a;
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H315-H319;
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.