Please login to your online account to display your discounted pricing

Iron(III) nitrate nonahydrate, 98+%, ACS reagent, ACROS Organics™


Quantity 10kg
Packaging Plastic Bucket

Manufacturer: thermo fisher scientific  AC423710100

 View more versions of this product

Catalog No. AC423710100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Adhering Crystalline Powder
Formula Weight 404
Color Light purple to grayish
Quantity 10kg
Merck Index 15, 4054
Packaging Plastic Bucket
Chemical Name or Material Iron(III) Nitrate Nonahydrate
UN Number 1466
Synonym Ferric Nitrate
Melting Point 47.0°C
Assay 98.0 to 101.0%
Decomposition Information Decomposition Information: < 100°C
Density 1.6800g/mL
Magnesium (Mg) 50ppm max.
Calcium (Ca) 100ppm max.
Insoluble Matter 0.005% max.
Potassium (K) 50ppm max.
Assay Percent Range 98+%
Sodium (Na) 500ppm max.
Molecular Formula FeN3O9·9H2O
CAS 7782-61-8
Additional Information Vapor Pressure: Negligible
Health Hazard 3 Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Irritating to eyes, respiratory system and skin. Hygroscopic.
NFPA
Health: 2
Flammability: 1
Instability: 2
Linear Formula Fe(NO3)3·9H2O
Sulfate 100ppm max.
Health Hazard 1 SIGNAL WORD: DANGER!
Health Hazard 2 GHS P Statement
Causes serious eye irritation.
Causes skin irritation.
May intensify fire; oxidizer.
Chlorine (Cl) 5ppm max.

Specifications

Chemical Name or Material Iron(III) Nitrate Nonahydrate
CAS 7782-61-8
Purity Grade ACS Reagent Grade
Assay 98.0 to 101.0%
Assay Percent Range 98+%
Molecular Formula FeN3O9·9H2O
Linear Formula Fe(NO3)3·9H2O
Formula Weight 404
Synonym Ferric Nitrate
Accessories

SIGNAL WORD: DANGER!

Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Irritating to eyes, respiratory system and skin. Hygroscopic.
NFPA
Health: 2
Flammability: 1
Instability: 2

GHS P Statement
Causes serious eye irritation.
Causes skin irritation.
May intensify fire; oxidizer.

SDS & Safety Documents