Learn More
Iron(III) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01071514
Description
It is used as catalyst for the synthesis of sodium amide from sodium and ammonia, and for etching of silver and silver alloys. Ferric nitrate is used in the synthesis of organically templated iron phosphates, and acts as an efficient catalyst in enamination of beta dicarbonyl compounds to beta enaminones. Ferric nitrate impregnated clays are useful as oxidizing agents. Clayfen, produced from ferric nitrate and montmorillonite, is used for the oxidation of thiols to disulfide, and alcohols to aldehydes. Ferric nitrate is useful for regio- and stereo-selective nitration of various aromatic, aliphatic, and heteroaromatic olefins. This reaction provides nitroolefins with excellent E-selectivity. With Keggin-type heteropoly acids, it selectively oxidizes various hydroxy groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
125°C (decomposition) | |
47°C | |
Purple | |
25 g | |
FeH18N3O18 | |
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate | |
SZQUEWJRBJDHSM-UHFFFAOYSA-N | |
iron(3+) nonahydrate trinitrate | |
16211566 | |
Acidic | |
Pale purple crystalline aggregates |
Iron(III) nitrate hydrate | |
7782-61-8 | |
Crystalline Aggregates | |
99.999% (Metal basis) | |
MFCD00149708 | |
Soluble in water,alcohol and acetone. | |
O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
403.99 | |
241.86 (Anhydrous) | |
Moisture sensitive | |
1.684 |
Chemical Identifiers
7782-61-8 | |
403.99 | |
SZQUEWJRBJDHSM-UHFFFAOYSA-N | |
16211566 | |
O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
FeH18N3O18 | |
MFCD00149708 | |
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate | |
iron(3+) nonahydrate trinitrate |
Safety and Handling
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
DOTInformation : Hazard Class: 5.1; Packaging Group: III
EINECSNumber : 233-899-5
RTECSNumber : NO7175000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only