Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(III) meso-tetraphenylporphine-mu-oxo dimer

Chemical Identifiers

Chemical Name or Material Iron(III) meso-tetraphenylporphine-mu-oxo dimer
CAS 12582-61-5
Formula Weight 1353.16
MDL Number MFCD00058898
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA33738ME

 
alfa aesar™
33738ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 1353.16
Chemical Name or Material Iron(III) meso-tetraphenylporphine-mu-oxo dimer
Physical Form Solid
MDL Number MFCD00058898
CAS 12582-61-5
Quantity 500mg