Learn More
Iron(III) isopropoxide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04487303
Description
Iron(III) isopropoxide is applied in the field of nano-materials and thin films, engineering and environmental field. It is employed in catalysis and synthesis, energy conversion and storage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Iron (III) isopropoxide | |
Gray | |
C9H21FeO3 | |
MFCD00070436 | |
UN3181 | |
iron 3+ ;propan-2-olate, iron iii i-propoxide, triisopropoxyiron iii, 2-propanol, iron 3+ salt 9ci, iron 3+ ion tris propan-2-olate, iron 3+ tris propan-2-olate, iron iii i-propoxide 99.9+%-fe | |
QUHDSMAREWXWFM-UHFFFAOYSA-N | |
iron(3+);propan-2-olate | |
11961763 | |
98% |
14995-22-3 | |
Ampule | |
Fe[OCH(CH3)2]3 | |
1 g | |
Moisture sensitive | |
Decomposes in water. | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Fe+3] | |
233.109 | |
233.11 | |
Solid Mass |
Chemical Identifiers
14995-22-3 | |
233.109 | |
QUHDSMAREWXWFM-UHFFFAOYSA-N | |
11961763 | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Fe+3] |
C9H21FeO3 | |
MFCD00070436 | |
iron 3+ ;propan-2-olate, iron iii i-propoxide, triisopropoxyiron iii, 2-propanol, iron 3+ salt 9ci, iron 3+ ion tris propan-2-olate, iron 3+ tris propan-2-olate, iron iii i-propoxide 99.9+%-fe | |
iron(3+);propan-2-olate |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA : No
storageNote1 :
- Ampouled under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only