Please login to your online account to display your discounted pricing

Iron(III) hydroxide, gamma-phase, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Iron(III) hydroxide
CAS 20344-49-4
Name Note gamma-phase
Molecular Formula FeO(OH)
Formula Weight 88.85
MDL Number MFCD00064782
Merck Index 14,4025
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1753104

  SDS
alfa aesar
1753104
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 88.85
Quantity 2g
Merck Index 14,4025
TSCA TSCA
Melting Point 135°C (Dec.)
Mesh Size −60 Mesh
Chemical Name or Material Iron(III) hydroxide
Odor Odorless
MDL Number MFCD00064782
CAS 20344-49-4
Physical Form Powder
Name Note gamma-phase
Molecular Formula FeO(OH)