missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(III) hydroxide, gamma-phase, Alfa Aesar™

Manufacturer:  Alfa Aesar 1753104

 View more versions of this product

Catalog No. AA1753104



Specifications

Specifications

Iron(III) hydroxide
20344-49-4
MFCD00064782
iron iii oxyhydroxide in water at ph = 3.0 +-0.5 feo-w320, iron iii oxide hydrated catalyst grade 30-50 mesh, iron iii oxide hydrated crystalline-325 mesh, iron hydroxide oxide fe oh o, iron pigment yellow e172, iron iii hydroxide gamma, iron iii hydroxide alpha, iron iii oxide-hydroxide, ferric hydroxide oxide
O[Fe]=O
88.851
88.85
−60 Mesh
135°C (decomposition)
gamma-phase
FeHO2
14,4025
AEIXRCIKZIZYPM-UHFFFAOYSA-M
hydroxy(oxo)iron
91502
Powder
Odorless
2g

Chemical Identifiers

20344-49-4
88.851
AEIXRCIKZIZYPM-UHFFFAOYSA-M
91502
O[Fe]=O
FeHO2
MFCD00064782
iron iii oxyhydroxide in water at ph = 3.0 +-0.5 feo-w320, iron iii oxide hydrated catalyst grade 30-50 mesh, iron iii oxide hydrated crystalline-325 mesh, iron hydroxide oxide fe oh o, iron pigment yellow e172, iron iii hydroxide gamma, iron iii hydroxide alpha, iron iii oxide-hydroxide, ferric hydroxide oxide
hydroxy(oxo)iron
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents