missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iron(III) hydroxide, alpha-phase, 99+%, Thermo Scientific Chemicals
$101.58 - $342.49
Chemical Identifiers
CAS | 20344-49-4 |
---|---|
Molecular Formula | FeHO2 |
Molecular Weight (g/mol) | 88.851 |
MDL Number | MFCD00064782 |
InChI Key | AEIXRCIKZIZYPM-UHFFFAOYSA-M |
Synonym | hydroxy oxo iron, goethite, lepidocrocite, iron hydroxide oxide, goethite fe oh o, iron hydroxide oxide fe oh o, iron iii oxide-hydroxide, ferric acid, iron pigment yellow, ferric hydroxide oxide |
PubChem CID | 91502 |
IUPAC Name | hydroxy(oxo)iron |
SMILES | O[Fe]=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1626730
|
Thermo Scientific Chemicals
A1626730 |
250 g |
Each for $101.58
|
|
|||||
AAA162670B
|
Thermo Scientific Chemicals
A162670B |
1000 g |
Each for $342.49
|
|
|||||
Description
Iron(III) hydroxide, alpha-phase, is used as a catalyst for proteomics research. It is also used as a pharamaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIron(III) hydroxide, alpha-phase, is used as a catalyst for proteomics research. It is also used as a pharamaceutical intermediate.
Solubility
It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
20344-49-4 | |
88.851 | |
AEIXRCIKZIZYPM-UHFFFAOYSA-M | |
91502 | |
O[Fe]=O |
FeHO2 | |
MFCD00064782 | |
hydroxy oxo iron, goethite, lepidocrocite, iron hydroxide oxide, goethite fe oh o, iron hydroxide oxide fe oh o, iron iii oxide-hydroxide, ferric acid, iron pigment yellow, ferric hydroxide oxide | |
hydroxy(oxo)iron |
Specifications
20344-49-4 | |
FeHO2 | |
MFCD00064782 | |
hydroxy oxo iron, goethite, lepidocrocite, iron hydroxide oxide, goethite fe oh o, iron hydroxide oxide fe oh o, iron iii oxide-hydroxide, ferric acid, iron pigment yellow, ferric hydroxide oxide | |
AEIXRCIKZIZYPM-UHFFFAOYSA-M | |
hydroxy(oxo)iron | |
91502 | |
≥99% | |
4.1 |
250 g | |
FeO(OH) | |
14,4025 | |
It is insoluble in water. | |
O[Fe]=O | |
88.851 | |
88.85 | |
Odorless | |
Iron(III) hydroxide, alpha-phase |
Safety and Handling
EINECSNumber : 243-746-4
RTECSNumber : NO7400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only