Alfa Aesar™ Iron(III) fluoride trihydrate
Manufacturer:  Alfa Aesar™ 1306818
 View more versions of this product
Catalog No. AA1306818
Specifications
Specifications
Iron(III) fluoride trihydrate
MFCD00149718
14,4022
166.89 (112.84 Anhydrous)
Hygroscopic
50g
15469-38-2
UN3260
iron 3+ trifluoride trihydrate, acmc-20akpl, ferric iron trihydrate trifluoride, iron 3+ ion trihydrate trifluoride, iron 3+ fluoride-water 1/3/3, fluoride, fluoride, iron salt, hydrate, hydrate, hydrate, fluoride
Crystalline
White to Brown
Chemical Identifiers
15469-38-2
MFCD00149718
iron 3+ trifluoride trihydrate, acmc-20akpl, ferric iron trihydrate trifluoride, iron 3+ ion trihydrate trifluoride, iron 3+ fluoride-water 1/3/3, fluoride, fluoride, iron salt, hydrate, hydrate, hydrate, fluoride
Safety and Handling
Safety and Handling
GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
TSCA : TSCA
Documents