missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(III) citrate hydrate, Fe(III) 16.5-20%; Fe(II) max 5%

Manufacturer:  Alfa Aesar™ A1027618

 View more versions of this product

Catalog No. AAA1027618



Description

Description

Applications
Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.

Solubility
Soluble in water.

Notes
Light sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Iron (III) citrate hydrate
C6H7FeO8
ferric citrate monohydrate, iron iii citrate hydrate, unii-334s8t6lke, ferric citrate hydrate, ferric citrate usan, iron iii citrate tribasic monohydrate, citric acid iron iii salt, iron cento citrate hydrate, ksc534c4h, iron 3+ citrate hydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[Fe+3]
262.959
244.94 (Anhydrous)
50g
334024-15-6
MFCD00149617
AJVRSHNXSHMMCH-UHFFFAOYSA-K
2-hydroxypropane-1,2,3-tricarboxylate;iron(3+);hydrate
22178220
Fe(III) 16.5 to 20%; Fe(II) max 5%

Chemical Identifiers

334024-15-6
262.959
AJVRSHNXSHMMCH-UHFFFAOYSA-K
22178220
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[Fe+3]
C6H7FeO8
MFCD00149617
ferric citrate monohydrate, iron iii citrate hydrate, unii-334s8t6lke, ferric citrate hydrate, ferric citrate usan, iron iii citrate tribasic monohydrate, citric acid iron iii salt, iron cento citrate hydrate, ksc534c4h, iron 3+ citrate hydrate
2-hydroxypropane-1,2,3-tricarboxylate;iron(3+);hydrate
SDS Documents