missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(Iii) Chloride Hexahydrate, puriss. p.a., Honeywell Fluka™
Puriss. p.a., Reagent European Pharmacopoeia,≥99%
Supplier: Honeywell-Fluka 3123225KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Iron(Iii) Chloride Hexahydrate | |
| 10025-77-1 | |
| Cl3FeH12O6 | |
| MFCD00149712 | |
| NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
| iron(3+) hexahydrate trichloride | |
| 270.3g/mol | |
| puriss. p.a. |
| Reag. Ph. Eur. | |
| 25 kg | |
| FeCl3 · 6H2O | |
| 3260 | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] | |
| 270.29 | |
| ≥99% | |
| 1mmHg (194°C) |
Chemical Identifiers
| 10025-77-1 | |
| 270.29 | |
| NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
| Cl3FeH12O6 | |
| MFCD00149712 | |
| iron(3+) hexahydrate trichloride |
Safety and Handling
P280-P305 + P351 + P338
H290-H302-H315-H318
EINECSNumber : 231-729-4
RTECSNumber : NO5425000