missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(III) chloride hexahydrate, ACS, 97.0-102.0%, Alfa Aesar

Manufacturer:  Alfa Aesar 1249730

 View more versions of this product

Catalog No. AA1249730



Description

Description

Applications:
  • Used to prepare organically templated Iron Phosphates
  • Used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin) process for production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards
  • Plays important role in photoengraving and photography as an etching agent
  • Usedl as mordant in dyeing and printing textiles.
  • Also usedl in detection of phenols and phenol derivatives; gamma-hydroxybutyric acids, in Trinder spot test for detecting salicylic acids
Specifications

Specifications

Iron(III) Chloride Hexahydrate
97.0 to 102.0%
MFCD00149712
Yellow-Brown lump
270.30 (162.21 Anhydrous)
ACS
Odorless
Brown-Yellow
10025-77-1
Cl3Fe·6H2O
UN3260
Ferric chloride hexahydrate
Lumps
1.820g/cm3
Freely soluble in water, alcohol, ether, acetone.Soluble in water, alcohol, glycerol, methanol, ether and acetone. Insoluble in ethyl acetate.
250g

Chemical Identifiers

10025-77-1
Cl3Fe·6H2O
MFCD00149712
Ferric chloride hexahydrate
Safety and Handling

Safety and Handling

H314-H318-H302: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.

EINECSNumber : 231-729-4

Documents