Learn More
Iron(III) chloride hexahydrate, ACS, 97.0-102.0%, Thermo Scientific Chemicals
Used to prepare organically templated iron phosphates.Also, used for sewerage treatment of industrial wastes, chloride hydrometallurgy. Useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.
Supplier: Thermo Scientific Chemicals 012497A7
Description
Iron(III) chloride hexahydrate is used to prepare organically templated iron phosphates. It is used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin process for the production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards. It plays an important role in photoengraving and photography as an etching agent. It is useful as a mordant in dyeing and printing textiles. It is useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications:- Used to prepare organically templated Iron Phosphates
- Used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin) process for production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards
- Plays important role in photoengraving and photography as an etching agent
- Usedl as mordant in dyeing and printing textiles.
- Also usedl in detection of phenols and phenol derivatives; gamma-hydroxybutyric acids, in Trinder spot test for detecting salicylic acids
Specifications
280°C to 285°C | |
37°C | |
Brown-Yellow | |
5 kg | |
Cl3FeH12O6 | |
UN3260 | |
Freely soluble in water, alcohol, ether, acetone.Soluble in water, alcohol, glycerol, methanol, ether and acetone. Insoluble in ethyl acetate. | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] | |
270.29 | |
ACS | |
Hygroscopic | |
1.820 g/cm3 |
Iron(III) Chloride Hexahydrate | |
10025-77-1 | |
Lump | |
97.0 to 102.0% | |
MFCD00149712 | |
Ferric chloride hexahydrate | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
iron(3+) hexahydrate trichloride | |
270.30 (162.21 Anhydrous) | |
Odorless | |
Yellow-Brown lump |
Chemical Identifiers
10025-77-1 | |
270.29 | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
iron(3+) hexahydrate trichloride |
Cl3FeH12O6 | |
MFCD00149712 | |
Ferric chloride hexahydrate | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
Safety and Handling
P260-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P314-P330-P333+P313-P362-P501c
H302-H315-H317-H318-H373
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 231-729-4
RTECSNumber : NO5425000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only