Learn More
Iron(III) chloride hexahydrate, 97+%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 423700100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
280.0°C to 285.0°C | |
37.0°C | |
White | |
10 kg | |
Cl3FeH12O6 | |
MFCD00149712 | |
15,4046 | |
Solubility in water: 920g/L (20°C). Other solubilities: readily soluble in alcohol, acetone and ether | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] | |
270.29 | |
≥97% | |
Plastic drum |
Iron(III) chloride hexahydrate | |
10025-77-1 | |
Crystalline Powder | |
97.0 to 102% (Iodometry) | |
FeCl3·6H2O | |
01,390; 02,199; 03,145; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 15,158 | |
Ferric chloride hexahydrate | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
iron(3+) hexahydrate trichloride | |
270.29 | |
ACS Reagent |
Chemical Identifiers
10025-77-1 | |
270.29 | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
iron(3+) hexahydrate trichloride |
Cl3FeH12O6 | |
MFCD00149712 | |
Ferric chloride hexahydrate | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with wa
GHS Signal Word: Danger
RTECSNumber : NO5425000
RUO – Research Use Only