missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(III) chloride, anhydrous, 98%, Alfa Aesar

Manufacturer:  Alfa Aesar 1235722*

 View more versions of this product

Catalog No. AA1235722



Specifications

Specifications

Iron(III) chloride
7705-08-0
Cl3Fe
Black crystalline
162.21
2.9g/cm3
Black
anhydrous
98%
MFCD00011005
Ferric chloride
Crystalline
Odorless
100g

Chemical Identifiers

7705-08-0
Cl3Fe
MFCD00011005
Ferric chloride
Safety and Handling

Safety and Handling

EINECSNumber : 231-729-4

storageNote1 : Hygroscopic

Documents