Learn More
Iron(III) chloride, 98%, pure, anhydrous, Thermo Scientific Chemicals
$67.91 - $191.53
Chemical Identifiers
CAS | 7705-08-0 |
---|---|
Molecular Formula | Cl3Fe |
Molecular Weight (g/mol) | 162.20 |
MDL Number | MFCD00011005 |
InChI Key | RBTARNINKXHZNM-UHFFFAOYSA-K |
Synonym | Ferric chloride |
IUPAC Name | iron(3+) trichloride |
SMILES | [Cl-].[Cl-].[Cl-].[Fe+3] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC169430050
|
Thermo Scientific Chemicals
169430050 |
5 g | Glass Bottle |
Each for $67.91
|
|
||||
AC169430010
|
Thermo Scientific Chemicals
169430010 |
1 kg | Plastic bottle |
Each for $99.60
|
|
||||
AC169430025
|
Thermo Scientific Chemicals
169430025 |
2.5 kg | Plastic bottle |
Each for $191.53
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
7705-08-0 | |
162.20 | |
RBTARNINKXHZNM-UHFFFAOYSA-K | |
iron(3+) trichloride |
Cl3Fe | |
MFCD00011005 | |
Ferric chloride | |
[Cl-].[Cl-].[Cl-].[Fe+3] |
Specifications
7705-08-0 | |
Powder | |
97.5 to 102.5% (Iodometry) | |
FeCl3 | |
01,392; 02,199; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 10,185; 11,237; 12,1; 13,133; 14,164; 16,167; 17,138 | |
Ferric chloride | |
RBTARNINKXHZNM-UHFFFAOYSA-K | |
iron(3+) trichloride | |
162.21 | |
Pure | |
Iron(III) chloride |
Gray to Black | |
5 g | |
Cl3Fe | |
MFCD00011005 | |
15,4046 | |
Solubility in water: 480g/L (20°C). Other solubilities: readily soluble in alcohol, ether and acetone, slightly soluble in cs2, practically insoluble in ethyl acetate | |
[Cl-].[Cl-].[Cl-].[Fe+3] | |
162.20 | |
98% | |
Glass Bottle |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
GHS Signal Word: Danger
EINECSNumber : 231-729-4
RUO – Research Use Only