missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(III) acetylacetonate, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 119130250

 View more versions of this product

Catalog No. AC119130250



Description

Description

Specifications

Specifications

Iron(III) acetylacetonate
Glass bottle
180.0°C to 183.0°C
99+%
99% min. (Iodometry)
MFCD00000020
2, 4-Pentanedione, iron(III) derivative, Ferric acetylacetonate
353.17
Powder
Authentic
Red
25g
14024-18-1
C15H21FeO6
04,268; 06,304; 13,211
Solubility in water: 2g/L (20°C). Other solubilities: readily soluble in acetone and chloroform,sparingly soluble in ether,40.1g/kg alcohol (20°C),535.4g/kg benzene (20°C)
353.17
99+%

Chemical Identifiers

14024-18-1
353.17
2, 4-Pentanedione, iron(III) derivative, Ferric acetylacetonate
C15H21FeO6
MFCD00000020
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 237-853-5

RTECSNumber : NO8960000

TSCA : TSCA

SDS Documents
Product Certifications