missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(III) 2-ethylhexanoate, nominally 50% in mineral spirits, Fe 6%

Manufacturer:  Alfa Aesar™ 89307A1

 View more versions of this product

Catalog No. AA89307A1



Specifications

Specifications

Iron (III) 2-ethylhexanoate
Brown
Nominally 50% in mineral spirits
C24H45FeO6
UN1268
Fe 6%
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Fe+3]
485.463
485.47
0.91g/mL
1kg
7321-53-1
MFCD00014006
ferric 2-ethylhexanoate, iron 2-ethylhexanoate, iron tris 2-ethylhexanoate, iron ethylhexanoate, iron iii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, iron 3+ salt 3:1, iron iii 2-ethylhexanoate nominally in mineral spirits, fe, iron tallate 2-ethylhexanoate, iron 3+ tris 2-ethylhexanoate, hexanoic acid, 2-ethyl-, iron 3+ salt
ULUYRVWYCIOFRV-UHFFFAOYSA-K
2-ethylhexanoate;iron(3+)
110963
Liquid

Chemical Identifiers

7321-53-1
485.463
ULUYRVWYCIOFRV-UHFFFAOYSA-K
110963
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Fe+3]
C24H45FeO6
MFCD00014006
ferric 2-ethylhexanoate, iron 2-ethylhexanoate, iron tris 2-ethylhexanoate, iron ethylhexanoate, iron iii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, iron 3+ salt 3:1, iron iii 2-ethylhexanoate nominally in mineral spirits, fe, iron tallate 2-ethylhexanoate, iron 3+ tris 2-ethylhexanoate, hexanoic acid, 2-ethyl-, iron 3+ salt
2-ethylhexanoate;iron(3+)
Safety and Handling

Safety and Handling

GHS H Statement
H340-H350-H372-H304-H226
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.

GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.

TSCA : TSCA

SDS Documents