Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(III) 2-ethylhexanoate, nominally 50% in mineral spirits, Fe 6%

Chemical Identifiers

Chemical Name or Material Iron (III) 2-ethylhexanoate
CAS 7321-53-1
Name Note Nominally 50% in mineral spirits
Molecular Formula C24H45FeO6
Formula Weight 485.47
MDL Number MFCD00014006
UN Number UN1268
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8930718

 
alfa aesar™
8930718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA8930730

 
alfa aesar™
8930730
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA89307A1

 
alfa aesar™
89307A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 485.47
TSCA TSCA
Name Note Nominally 50% in mineral spirits
Physical Form Liquid
Density 0.91g/mL
Molecular Formula C24H45FeO6
MDL Number MFCD00014006
CAS 7321-53-1
UN Number UN1268
Color Brown
Quantity 50g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H340-H350-H372-H304-H226
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.
Chemical Name or Material Iron (III) 2-ethylhexanoate
Concentration Fe 6%
Safety and Handling

Danger

GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H340-H350-H372-H304-H226
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.