Learn More
Iron(III) 2,4-pentanedionate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01253422
Description
Iron(III) 2,4-pentanedionate is used as a precatalyst and reagent in organic chemistry. It acts as a precursor for highly crystalline (Zn, Fe)Fe2O4 films and magnetic property measurements of these films. It catalyzes the dimerization of isoprene to a mixture of 1,5-dimethyl-1,5-cyclooctadiene and 2,5-dimethyl-1,5-cyclooctadiene, ring-opening polymerization of 1,3-benzoxazine and catalyze the reaction of N-sulfonyl oxaziridines with olefins to form 1,3-oxazolidine products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Iron(III) 2,4-pentanedionate | |
∼183°C (decomposition) | |
C15H21FeO6 | |
100 g | |
Iron(III) acetylacetonate; Tris(acetylacetonato)iron(III) | |
AQBLLJNPHDIAPN-LNTINUHCSA-K | |
iron(3+) tris((2Z)-4-oxopent-2-en-2-olate) | |
353.17 |
14024-18-1 | |
Red | |
MFCD00000020 | |
4157960 | |
Soluble in toluene. | |
[Fe+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
353.17 | |
Powder |
Chemical Identifiers
14024-18-1 | |
353.17 | |
AQBLLJNPHDIAPN-LNTINUHCSA-K | |
iron(3+) tris((2Z)-4-oxopent-2-en-2-olate) |
C15H21FeO6 | |
MFCD00000020 | |
Iron(III) acetylacetonate; Tris(acetylacetonato)iron(III) | |
[Fe+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Safety and Handling
GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
H302-H319-H335-H500
EINECSNumber : 237-853-5
RTECSNumber : NO8960000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only