Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(III) 1,3-diphenyl-1,3-propanedionate

Chemical Identifiers

Chemical Name or Material Iron(III) 1,3-diphenyl-1,3-propanedionate
Molecular Formula C45H33FeO6
Formula Weight 725.61
MDL Number MFCD14636458
Synonym Iron(III) (DBM)3; Tris(dibenzoylmethanato)iron(III)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3227806

  SDS
alfa aesar™
H3227806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 725.61
Chemical Name or Material Iron(III) 1,3-diphenyl-1,3-propanedionate
MDL Number MFCD14636458
Synonym Iron(III) (DBM)3; Tris(dibenzoylmethanato)iron(III)
Molecular Formula C45H33FeO6
Quantity 5g