missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iron(II) sulfide, fused sticks, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific 233300010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Iron(II) sulfide | |
1317-37-9 | |
1195.0°C | |
FeS | |
MFCD00011013 | |
ferrous monosulfide, ferrous sulfide, iron ii sulfide, iron monosulfide, iron protosulfide, iron sulfide, iron sulfuret, iron sulphide, troilite | |
MBMLMWLHJBBADN-UHFFFAOYSA-N | |
sulfanylideneiron | |
14828 | |
Sticks | |
Plastic bottle | |
4.84 |
c.p., 60-67% Fe | |
Black-Gray | |
1kg | |
FeS | |
15, 4085 | |
Solubility in water: practically insoluble. Other solubilities: soluble in acids with the evolution of h2s | |
S=[Fe] | |
87.91 | |
87.92 | |
≈25% (S) | |
Solubility in water: practically insoluble. Other solubilities: soluble in acids with the evolution of h2s | |
4.8400g/mL |
Safety and Handling
MOISTURE SENSITIVE
EINECSNumber : 215-268-6
TSCA : TSCA
RUO – Research Use Only