Please login to your online account to display your discounted pricing

Iron(II) sulfide, fused sticks, ACROS Organics™

$129.10 - $129.10

Chemical Identifiers

Chemical Name or Material Iron(II) sulfide
Name Note c.p., 60-67% Fe
CAS 1317-37-9
Assay ≈25% (S)
Molecular Formula FeS
Linear Formula FeS
Formula Weight 87.92
MDL Number MFCD00011013
Synonym Ferrous sulfide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC233300010

  SDS
thermo fisher scientific
233300010
Plastic bottle 1kg Each for $129.10
Description & Specifications

Specifications

Assay ≈25% (S)
Color Grayish-black
Physical Form Sticks
Formula Weight 87.92
Quantity 1kg
TSCA TSCA
EINECS Number 215-268-6
Density 4.8400g/mL
Merck Index 15, 4085
Linear Formula FeS
Packaging Plastic bottle
Name Note c.p., 60-67% Fe
MDL Number MFCD00011013
Chemical Name or Material Iron(II) sulfide
CAS 1317-37-9
Molecular Formula FeS
Synonym Ferrous sulfide
Health Hazard 1 MOISTURE SENSITIVE
Safety and Handling

MOISTURE SENSITIVE