missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(II) sulfide, 99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1742218

 View more versions of this product

Catalog No. AA1742218



Specifications

Specifications

Iron(II) sulfide
99% (Metal basis)
MFCD00011013
MBMLMWLHJBBADN-UHFFFAOYSA-N
sulfanylideneiron
14828
Powder
Brown to Black
1317-37-9
FeS
Brown to black powder
S=[Fe]
87.905
87.91
Odorless
50g

Chemical Identifiers

1317-37-9
87.905
MBMLMWLHJBBADN-UHFFFAOYSA-N
FeS
MFCD00011013
14828
Safety and Handling

Safety and Handling

EINECSNumber : 215-268-6

SDS Documents