missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(II) sulfide, 99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1742209

 View more versions of this product

Catalog No. AA1742209



Specifications

Specifications

Iron(II) sulfide
99% (Metal basis)
MFCD00011013
ferrous sulfide, iron ii sulfide, iron sulfide, iron ii sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
S=[Fe]
87.905
87.91
4.74g/cm3
Brown to Black
1317-37-9
FeS
Brown to black powder
MBMLMWLHJBBADN-UHFFFAOYSA-N
sulfanylideneiron
14828
Powder
Odorless
10g

Chemical Identifiers

1317-37-9
87.905
MBMLMWLHJBBADN-UHFFFAOYSA-N
14828
S=[Fe]
FeS
MFCD00011013
ferrous sulfide, iron ii sulfide, iron sulfide, iron ii sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
sulfanylideneiron
Safety and Handling

Safety and Handling

EINECSNumber : 215-268-6

SDS Documents