missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(II) sulfide, 99.98% (metals basis)

Manufacturer:  Alfa Aesar™ 2238806

 View more versions of this product

Catalog No. AA2238806



Specifications

Specifications

Iron(II) sulfide
99.98%
FeS
14,4058
S=[Fe]
87.905
87.91
4.74g/mL
Moisture Sensitive
1195°C
1317-37-9
(metals basis)
MFCD00011013
MBMLMWLHJBBADN-UHFFFAOYSA-N
sulfanylideneiron
14828
Powder
Odorless
Brown to Black
5g

Chemical Identifiers

1317-37-9
87.905
MBMLMWLHJBBADN-UHFFFAOYSA-N
sulfanylideneiron
FeS
MFCD00011013
14828
S=[Fe]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents