missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(II) sulfide, 99.98% (metals basis)

Manufacturer:  Alfa Aesar™ 2238814

 View more versions of this product

Catalog No. AA2238814



Specifications

Specifications

Iron(II) sulfide
(metals basis)
MFCD00011013
ferrous sulfide, iron ii sulfide, iron sulfide, iron ii sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
S=[Fe]
87.905
87.91
99.98%
Odorless
Brown to Black
25g
1317-37-9
FeS
14,4058
MBMLMWLHJBBADN-UHFFFAOYSA-N
sulfanylideneiron
14828
Powder
4.74g/mL
Moisture Sensitive
1195°C

Chemical Identifiers

1317-37-9
87.905
MBMLMWLHJBBADN-UHFFFAOYSA-N
14828
S=[Fe]
FeS
MFCD00011013
ferrous sulfide, iron ii sulfide, iron sulfide, iron ii sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
sulfanylideneiron
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents