missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1296714

 View more versions of this product

Catalog No. AA1296714



Specifications

Specifications

Iron(II) sulfate heptahydrate
(metals basis)
FeSO4·7H2O
14,4057
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron(2+);sulfate;heptahydrate
62662
278.02 (151.91 Anhydrous)
99.999%
1.897g/mL
Green
25g
7782-63-0
FeH14O11S
MFCD00149719
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Fe+2]
278.006
CHEBI:75836
Crystalline
Puratronic™
Air Sensitive
64°C (in -6H2O)

Chemical Identifiers

7782-63-0
278.006
SURQXAFEQWPFPV-UHFFFAOYSA-L
62662
iron(2+);sulfate;heptahydrate
FeH14O11S
MFCD00149719
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
CHEBI:75836
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Fe+2]
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see ... on this label).Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NO8510000

TSCA : TSCA

Documents