Learn More
Iron(II) perchlorate hydrate, Reagent Grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01164636
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Iron(II) perchlorate hydrate | |
335159-18-7 | |
500 g | |
MFCD00150234 | |
Soluble in alcohol and water (?98g/100 mL H2O at 0?°C). | |
[Fe++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O | |
254.74 | |
Reagent |
>100°C (decomposition) | |
Crystalline | |
Cl2FeO8 | |
UN1481 | |
CKFMJXZQTNRXGX-UHFFFAOYSA-L | |
λ²-iron(2+) diperchlorate | |
254.75 (Anhydrous) | |
Hygroscopic |
Chemical Identifiers
335159-18-7 | |
254.74 | |
CKFMJXZQTNRXGX-UHFFFAOYSA-L | |
[Fe++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O |
Cl2FeO8 | |
MFCD00150234 | |
λ²-iron(2+) diperchlorate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber : 237-704-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only