missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(II) oxide, 99.5% (metals basis), Alfa Aesar

In manufacture of green, heat-absorbing glass

Manufacturer:  Alfa Aesar 3051314

 View more versions of this product

Catalog No. AA3051314



Specifications

Specifications

Iron(II) oxide
99.5% (metals basis)
iron ii oxide, ferrous oxide, iron monooxide, iron monoxide, iron oxide, wustite, natural wuestite, eisenoxydul, hsdb 464, eisen ii-oxid
O=[Fe]
71.844
CHEBI:50820
5 Mesh
25g
1345-25-1
FeO
UQSXHKLRYXJYBZ-UHFFFAOYSA-N
oxoiron
14945
71.85
5.7

Chemical Identifiers

1345-25-1
71.844
iron ii oxide, ferrous oxide, iron monooxide, iron monoxide, iron oxide, wustite, natural wuestite, eisenoxydul, hsdb 464, eisen ii-oxid
CHEBI:50820
O=[Fe]
FeO
UQSXHKLRYXJYBZ-UHFFFAOYSA-N
14945
oxoiron
Safety and Handling

Safety and Handling

storageNote1 : Air Sensitive

SDS Documents