missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(II) methoxide, 98%

Manufacturer:  Alfa Aesar™ 2255204

 View more versions of this product

Catalog No. AA2255204



Description

Description

  • Packaged under argon
Specifications

Specifications

Iron(II) methoxide
Air & Moisture Sensitive
2g
C2H6FeO2
MFCD00061474
iron ii methoxide, iron 2+ dimethoxide, methanol, iron 2+ salt, acmc-1bc40, iron ii methoxide
C[O-].C[O-].[Fe+2]
117.913
117.92
98%
Alcohol-like
Green
7245-21-8
Fe(OCH3)2
UN3181
VAPRHKOWFRYFTF-UHFFFAOYSA-N
iron(2+);methanolate
14598007
Powder

Chemical Identifiers

7245-21-8
117.913
VAPRHKOWFRYFTF-UHFFFAOYSA-N
14598007
C[O-].C[O-].[Fe+2]
C2H6FeO2
MFCD00061474
iron ii methoxide, iron 2+ dimethoxide, methanol, iron 2+ salt, acmc-1bc40, iron ii methoxide
iron(2+);methanolate
Safety and Handling

Safety and Handling

GHS H Statement
H228-H302-H315-H319-H335
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P280h-P305+P351+P338-P337+P313
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

storageNote1 :

  • Packaged under argon

SDS Documents