Learn More
Iron(II) methoxide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 02255209
Description
Iron(II) methoxide, is used in Engineering & Environmental, Catalysis & Synthesis, and In organic syntheses, chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Iron(II) methoxide | |
Green | |
C2H6FeO2 | |
MFCD00061474 | |
UN3181 | |
iron ii methoxide, iron 2+ dimethoxide, methanol, iron 2+ salt, acmc-1bc40 | |
VAPRHKOWFRYFTF-UHFFFAOYSA-N | |
iron(2+);methanolate | |
14598007 | |
98% |
7245-21-8 | |
Alcohol-like | |
Fe(OCH3)2 | |
10 g | |
Air and moisture sensitive | |
Reacts with water. | |
C[O-].C[O-].[Fe+2] | |
117.913 | |
117.92 | |
Powder |
Chemical Identifiers
7245-21-8 | |
117.913 | |
VAPRHKOWFRYFTF-UHFFFAOYSA-N | |
14598007 | |
C[O-].C[O-].[Fe+2] |
C2H6FeO2 | |
MFCD00061474 | |
iron ii methoxide, iron 2+ dimethoxide, methanol, iron 2+ salt, acmc-1bc40 | |
iron(2+);methanolate |
Safety and Handling
GHS H Statement
H228-H302-H315-H319-H335
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA : No
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only