missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(II) iodide, ultra dry, 99.99% (metals basis), Alfa Aesar™

Light and Air Sensitive and Hygroscopic

Manufacturer:  Alfa Aesar 1361909

 View more versions of this product

Catalog No. AA1361909



Specifications

Specifications

Iron(II) iodide
99.99% (Metal basis)
MFCD00016094
iron iodide, iron ii iodide, iron ii iodide, acmc-1bdqb, iron ii iodide, anhydrous, iron ii iodide, ultra dry, iron ii iodide, anhydrous, beads,-10 mesh trace metals basis
Beads
Gray
7783-86-0
FeI2
Gray beads
309.66
Odorless
10g

Chemical Identifiers

7783-86-0
FeI2
MFCD00016094
iron iodide, iron ii iodide, iron ii iodide, acmc-1bdqb, iron ii iodide, anhydrous, iron ii iodide, ultra dry, iron ii iodide, anhydrous, beads,-10 mesh trace metals basis
Safety and Handling

Safety and Handling

H314-H318-H302: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.

EINECSNumber : 232-031-2

Documents