missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(II) iodide, anhydrous, 97% (metals basis)

Manufacturer:  Alfa Aesar™ 1148504*

 View more versions of this product

Catalog No. AA1148504



Specifications

Specifications

Iron(II) iodide
7783-86-0
MFCD00016094
iron iodide, iron ii iodide, iron ii iodide, acmc-1bdqb, iron ii iodide, anhydrous, iron ii iodide, ultra dry, iron ii iodide, anhydrous, beads,-10 mesh trace metals basis
Crystalline
5.32g/mL
Light & Air Sensitive & Hygroscopic
2g
Anhydrous
(metals basis)
UN3260
309.66
97%
Odorless
Brown

Chemical Identifiers

7783-86-0
MFCD00016094
iron iodide, iron ii iodide, iron ii iodide, acmc-1bdqb, iron ii iodide, anhydrous, iron ii iodide, ultra dry, iron ii iodide, anhydrous, beads,-10 mesh trace metals basis
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents