Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(II) iodide, anhydrous, 97% (metals basis)

$79.41 - $79.41

Chemical Identifiers

Chemical Name or Material Iron(II) iodide
CAS 7783-86-0
Name Note Anhydrous
Formula Weight 309.66
MDL Number MFCD00016094
UN Number UN3260
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1148504

  SDS
alfa aesar™
1148504*
2g Each for $79.41

AA1148509

  SDS
alfa aesar™
1148509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 309.66
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Chemical Name or Material Iron(II) iodide
TSCA TSCA
Odor Odorless
Name Note Anhydrous
Physical Form Crystalline
Assay 97%
Sensitivity Light & Air Sensitive & Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00016094
CAS 7783-86-0
UN Number UN3260
Color Brown
Quantity 2g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Density 5.32g/mL
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.