Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(II,III) oxide, Puratronic™, 99.997% (metals basis)

$210.95 - $210.95

Chemical Identifiers

Chemical Name or Material Iron(II,III) oxide
CAS 1317-61-9
Purity Grade Puratronic™
Formula Weight 231.54
MDL Number MFCD00011010
Merck Index 14,4040
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1296214

  SDS
alfa aesar™
1296214
25g Each for $210.95

AA1296222

  SDS
alfa aesar™
1296222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 231.54
Quantity 25g
Color Black
Odor Odorless
Purity Grade Puratronic™
MDL Number MFCD00011010
Melting Point 1538°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Assay 99.997%
CAS 1317-61-9
Physical Form Powder
Merck Index 14,4040
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
Chemical Name or Material Iron(II,III) oxide
Density 5.2g/mL
Refractive Index 3.0
Assay Percent Notes (metals basis)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.