missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(II) gluconate hydrate, 97%

Manufacturer:  Alfa Aesar™ 88726A3

 View more versions of this product

Catalog No. AA88726A3



Specifications

Specifications

Iron (II) gluconate hydrate
97%
MFCD00150872
2kg
C(C(C(C(C(C(=O)O)O)O)O)O)O.C(C(C(C(C(C(=O)O)O)O)O)O)O.O.O.[Fe]
484.185
446.19 (Anhydrous)
22830-45-1
C12H28FeO16
~140°C
DLTLQHHLCQRGHU-SYAJEJNSSA-N
iron;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;dihydrate
131851469
Granular

Chemical Identifiers

22830-45-1
484.185
DLTLQHHLCQRGHU-SYAJEJNSSA-N
iron;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;dihydrate
C12H28FeO16
MFCD00150872
131851469
C(C(C(C(C(C(=O)O)O)O)O)O)O.C(C(C(C(C(C(=O)O)O)O)O)O)O.O.O.[Fe]
Safety and Handling

Safety and Handling

RTECSNumber : LZ5180000

TSCA : TSCA

SDS Documents