missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(II) fluoride, anhydrous, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01148609

 View more versions of this product

Catalog No. AA1148609



Description

Description

Iron(II) fluoride is used for proteomics research applications. It is also a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Iron(II) fluoride
Anhydrous
White
10 g
MFCD00016093
14,4045
FZGIHSNZYGFUGM-UHFFFAOYSA-L
λ²-iron(2+) difluoride
93.84
Air sensitive; Hygroscopic
>1000°C (sublimation)
7789-28-8
Powder
F2Fe
UN3260
Insoluble in water.
[F-].[F-].[Fe++]
93.84
98%
4.09 g/mL

Chemical Identifiers

7789-28-8
93.84
FZGIHSNZYGFUGM-UHFFFAOYSA-L
[F-].[F-].[Fe++]
F2Fe
MFCD00016093
λ²-iron(2+) difluoride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 232-155-7

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only