missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iron(II) ethylenediammonium sulfate tetrahydrate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03054722
Description
Iron(II) ethylenediammonium sulfate tetrahydrate is used in the preparation of gold nanoparticles by reacting with gold(III)-bis[ethylene-diamine]-chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Iron(II) ethylenediammonium sulfate tetrahydrate | |
Green to White | |
MFCD00082474 | |
Soluble in water. | |
99% |
113193-60-5 | |
FeC2H4(NH3)2(SO4)2·4H2O | |
100 g | |
382.14 (310.08 Anhydrous) | |
Crystalline |
Chemical Identifiers
113193-60-5 | |
MFCD00082474 |
Safety and Handling
EINECSNumber : 264-357-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only