missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(II) ethylenediammonium sulfate tetrahydrate, 99%

Manufacturer:  Alfa Aesar™ 3054714

 View more versions of this product

Catalog No. AA3054714



Specifications

Specifications

Iron(II) ethylenediammonium sulfate tetrahydrate
25g
FeC2H4(NH3)2(SO4)2·4H2O
382.14 (310.08 Anhydrous)
99%
Green to White
34962-29-3
MFCD00082474
Crystalline

Chemical Identifiers

34962-29-3
MFCD00082474
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents