Please login to your online account to display your discounted pricing

Iron(II) chloride, anhydrous, 99.5% (metals basis), Alfa Aesar™

$162.73 - $162.73

Chemical Identifiers

Chemical Name or Material Iron(II) chloride, anhydrous
CAS 7758-94-3
Physical Form Powder
Assay Percent Range 99.5% (Metal basis)
Molecular Formula Cl2Fe
Formula Weight 126.75
MDL Number MFCD00011004
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3114118

  SDS
alfa aesar
3114118
50g Each for $162.73

AA3114130

  SDS
alfa aesar
3114130
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.75
Quantity 50g
Color Off-white
DOT Information DOT Class: 8, Packing Group: III
Physical Form Powder
EINECS Number 231-843-4
MDL Number MFCD00011004
Molecular Formula Cl2Fe
Density 3.16g/cm3
CAS 7758-94-3
Health Hazard 1 H314-H318-H302: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.
Appearance Off-white powder
Odor Odorless
Assay Percent Range 99.5% (Metal basis)
Chemical Name or Material Iron(II) chloride, anhydrous
Safety and Handling

H314-H318-H302: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.